Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCAGGGGTGACCCAGAGCACCAT[A/G]TTTTCCTGTGAAGCTCACAACCTAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600341 | ||||||||||||||||||||
Literature Links: |
TYRO3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TYRO3 - TYRO3 protein tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006293.3 | 637 | Missense Mutation | ATA,ATG | I,M 200 | NP_006284.2 | |
XM_017022542.1 | 637 | Missense Mutation | ATA,ATG | I,M 155 | XP_016878031.1 | |
XM_017022543.1 | 637 | Missense Mutation | ATA,ATG | I,M 200 | XP_016878032.1 |