Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCCAGCATCGCCAGACTCCGCAT[C/T]GCAAAGGGGGGCGTCAATGACAATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 188060 | ||||||||||||||||||||
Literature Links: |
THBS1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
THBS1 - thrombospondin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003246.3 | 772 | Silent Mutation | ATC,ATT | I,I 199 | NP_003237.2 | |
XM_011521971.1 | 772 | Silent Mutation | ATC,ATT | I,I 199 | XP_011520273.1 |