Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAACAAATCTCAATTGTGAGTTCCT[C/T]GCAATATCGTTCAGGCTACTAAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTXN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CTXN2 - cortexin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145668.1 | Intron | NP_001139140.1 | ||||
XM_005254378.3 | Intron | XP_005254435.1 | ||||
XM_006720504.2 | Intron | XP_006720567.1 | ||||
XM_011521553.1 | Intron | XP_011519855.1 | ||||
XM_017022178.1 | Intron | XP_016877667.1 |
MYEF2 - myelin expression factor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |