Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCTGCCCTCAGGGCTGGGCACCA[C/T]TGGCACCCAACAGGGCCGTCTTGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602059 MIM: 610745 | ||||||||||||||||||||
Literature Links: |
ISLR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ISLR - immunoglobulin superfamily containing leucine rich repeat | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STRA6 - stimulated by retinoic acid 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142617.1 | 2200 | Missense Mutation | AAT,AGT | N,S 663 | NP_001136089.1 | |
NM_001142618.1 | 2200 | Missense Mutation | AAT,AGT | N,S 663 | NP_001136090.1 | |
NM_001142619.1 | 2200 | Missense Mutation | AAT,AGT | N,S 654 | NP_001136091.1 | |
NM_001142620.1 | 2200 | Intron | NP_001136092.1 | |||
NM_001199040.1 | 2200 | Missense Mutation | AAT,AGT | N,S 700 | NP_001185969.1 | |
NM_001199041.1 | 2200 | Missense Mutation | AAT,AGT | N,S 678 | NP_001185970.1 | |
NM_001199042.1 | 2200 | Missense Mutation | AAT,AGT | N,S 702 | NP_001185971.1 | |
NM_022369.3 | 2200 | Missense Mutation | AAT,AGT | N,S 663 | NP_071764.3 | |
XM_011521883.1 | 2200 | Missense Mutation | AAT,AGT | N,S 663 | XP_011520185.1 | |
XM_011521884.1 | 2200 | Missense Mutation | AAT,AGT | N,S 600 | XP_011520186.1 | |
XM_011521885.2 | 2200 | Intron | XP_011520187.1 | |||
XM_017022478.1 | 2200 | Missense Mutation | AAT,AGT | N,S 679 | XP_016877967.1 | |
XM_017022479.1 | 2200 | Missense Mutation | AAT,AGT | N,S 663 | XP_016877968.1 | |
XM_017022480.1 | 2200 | Missense Mutation | AAT,AGT | N,S 600 | XP_016877969.1 |