Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATGTGGTAGGGCCGGGCCGCCGC[A/G]TTCATGTTGACCCTCCGGATCCCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614362 MIM: 601149 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACSBG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACSBG1 - acyl-CoA synthetase bubblegum family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199377.1 | 2207 | Silent Mutation | AAC,AAT | N,N 667 | NP_001186306.1 | |
NM_015162.4 | 2207 | Silent Mutation | AAC,AAT | N,N 671 | NP_055977.3 | |
XM_011521390.2 | 2207 | Silent Mutation | AAC,AAT | N,N 649 | XP_011519692.1 | |
XM_011521391.2 | 2207 | Silent Mutation | AAC,AAT | N,N 600 | XP_011519693.2 | |
XM_017022025.1 | 2207 | Silent Mutation | AAC,AAT | N,N 671 | XP_016877514.1 | |
XM_017022026.1 | 2207 | Silent Mutation | AAC,AAT | N,N 679 | XP_016877515.1 |
IDH3A - isocitrate dehydrogenase 3 (NAD(+)) alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |