Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGACTATTCACATGAAACTGCGTC[C/G]ACCGACGATGCTGGAATCCAATATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608689 | ||||||||||||||||||||
Literature Links: |
MESP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MESP1 - mesoderm posterior bHLH transcription factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018670.3 | 1814 | Intron | NP_061140.1 |
WDR93 - WD repeat domain 93 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284395.1 | 1814 | Silent Mutation | TCC,TCG | S,S 569 | NP_001271324.1 | |
NM_001284396.1 | 1814 | Intron | NP_001271325.1 | |||
NM_020212.1 | 1814 | Silent Mutation | TCC,TCG | S,S 597 | NP_064597.1 | |
XM_006720614.1 | 1814 | Missense Mutation | CCA,CGA | P,R 576 | XP_006720677.1 | |
XM_006720615.1 | 1814 | Missense Mutation | CCA,CGA | P,R 547 | XP_006720678.1 | |
XM_006720618.2 | 1814 | Intron | XP_006720681.1 | |||
XM_011521794.2 | 1814 | Missense Mutation | CCA,CGA | P,R 576 | XP_011520096.1 | |
XM_011521795.1 | 1814 | Missense Mutation | CCA,CGA | P,R 576 | XP_011520097.1 | |
XM_011521796.2 | 1814 | Intron | XP_011520098.1 | |||
XM_011521798.2 | 1814 | Missense Mutation | CCA,CGA | P,R 378 | XP_011520100.1 | |
XM_011521800.1 | 1814 | Missense Mutation | CCA,CGA | P,R 378 | XP_011520102.1 | |
XM_011521802.2 | 1814 | Intron | XP_011520104.1 | |||
XM_017022415.1 | 1814 | Missense Mutation | CCA,CGA | P,R 400 | XP_016877904.1 | |
XM_017022416.1 | 1814 | Missense Mutation | CCA,CGA | P,R 378 | XP_016877905.1 | |
XM_017022417.1 | 1814 | Intron | XP_016877906.1 |