Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTCATCTGCCAGCAGCCATGTTT[C/T]CTGCAGCATACTCCTGCGGGCAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 151520 MIM: 611475 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LTK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LTK - leukocyte receptor tyrosine kinase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPAP1 - RNA polymerase II associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015540.3 | 4423 | Missense Mutation | AAA,GAA | K,E 1337 | NP_056355.2 | |
XM_005254297.1 | 4423 | Missense Mutation | AAA,GAA | K,E 1337 | XP_005254354.1 | |
XM_006720469.1 | 4423 | Missense Mutation | AAA,GAA | K,E 1277 | XP_006720532.1 |