Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAGGCAGGCTCAGAGCTCGCAAG[C/G]CTCCCACGTGTTCCCCAAAGAGGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 151520 MIM: 611475 | ||||||||||||||||||||
Literature Links: |
LTK PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LTK - leukocyte receptor tyrosine kinase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPAP1 - RNA polymerase II associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015540.3 | 4178 | Missense Mutation | GCC,GGC | A,G 1255 | NP_056355.2 | |
XM_005254297.1 | 4178 | Missense Mutation | GCC,GGC | A,G 1255 | XP_005254354.1 | |
XM_006720469.1 | 4178 | Missense Mutation | GCC,GGC | A,G 1195 | XP_006720532.1 |