Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTGGGCTGCCCAGAGGCCTGAAG[A/G]GCATGCTGTAGGGCTTGGCTGAAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609748 | ||||||||||||||||||||
Literature Links: |
UBL7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
UBL7 - ubiquitin like 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286739.1 | 1085 | Silent Mutation | GCC,GCT | A,A 325 | NP_001273668.1 | |
NM_001286740.1 | 1085 | Silent Mutation | GCC,GCT | A,A 325 | NP_001273669.1 | |
NM_001286741.1 | 1085 | Silent Mutation | GCC,GCT | A,A 365 | NP_001273670.1 | |
NM_001286742.1 | 1085 | Silent Mutation | GCC,GCT | A,A 365 | NP_001273671.1 | |
NM_032907.4 | 1085 | Silent Mutation | GCC,GCT | A,A 325 | NP_116296.1 | |
NM_201265.1 | 1085 | Silent Mutation | GCC,GCT | A,A 325 | NP_957717.1 | |
XM_011522124.2 | 1085 | Silent Mutation | GCC,GCT | A,A 365 | XP_011520426.1 |