Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACGGGCATGGCCCGGGCCCGCAC[C/T]TCCGTCTCCCAGCAACGGCACGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 153456 MIM: 616800 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOXL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOXL1 - lysyl oxidase like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005576.3 | 765 | Silent Mutation | ACC,ACT | T,T 144 | NP_005567.2 | |
XM_011521555.1 | 765 | Silent Mutation | ACC,ACT | T,T 144 | XP_011519857.1 | |
XM_017022179.1 | 765 | Intron | XP_016877668.1 |
LOXL1-AS1 - LOXL1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |