Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGCAGGTTGCCCCTTTTTCATCA[G/T]CACCAGAACAACAAATGTAAGTATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608238 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPPL2A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPPL2A - signal peptide peptidase like 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032802.3 | 1549 | Missense Mutation | ATG,CTG | M,L 457 | NP_116191.2 | |
XM_005254722.3 | 1549 | Missense Mutation | ATG,CTG | M,L 475 | XP_005254779.1 | |
XM_017022680.1 | 1549 | Intron | XP_016878169.1 | |||
XM_017022681.1 | 1549 | Intron | XP_016878170.1 |