Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTGAGGGTGACATGGACTCTCAG[C/T]GCCTTGCCCACCTCAGCCCTCTCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606776 | ||||||||||||||||||||
Literature Links: |
TGM7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TGM7 - transglutaminase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052955.2 | 1872 | Silent Mutation | GCA,GCG | A,A 623 | NP_443187.1 | |
XM_017021903.1 | 1872 | Silent Mutation | GCA,GCG | A,A 624 | XP_016877392.1 |