Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATGGGAGCTGCAGAGTTGTGCTG[C/T]GAGTCATTGCTTCAAACAAGCTCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
MIR3174 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR3174 - microRNA 3174 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF710 - zinc finger protein 710 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198526.3 | Intron | NP_940928.2 | ||||
XM_005254906.3 | Intron | XP_005254963.1 | ||||
XM_005254907.3 | Intron | XP_005254964.1 | ||||
XM_011521540.2 | Intron | XP_011519842.1 | ||||
XM_011521541.2 | Intron | XP_011519843.1 | ||||
XM_011521542.2 | Intron | XP_011519844.1 | ||||
XM_011521543.2 | Intron | XP_011519845.1 | ||||
XM_011521544.1 | Intron | XP_011519846.1 |