Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAATGGAGAAATTGGATGTGATC[A/G]GATAGTCACTGGGAACTTTATATTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609096 MIM: 610894 | ||||||||||||||||||||
Literature Links: |
FBXO22 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FBXO22 - F-box protein 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012170.3 | 1205 | Intron | NP_036302.1 | |||
NM_147188.2 | 1205 | Missense Mutation | CAG,CGG | Q,R 367 | NP_671717.1 | |
XM_011521447.2 | 1205 | Missense Mutation | CAG,CGG | Q,R 263 | XP_011519749.1 |
FBXO22-AS1 - FBXO22 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NRG4 - neuregulin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138573.3 | 1205 | Intron | NP_612640.1 |