Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCATCTCTAGCTCAGCGATCCGCC[A/G]TCCCAGACACTGCCGCACACCCCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 118485 | ||||||||||||||||||||
Literature Links: |
CCDC33 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC33 - coiled-coil domain containing 33 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CYP11A1 - cytochrome P450 family 11 subfamily A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000781.2 | 1622 | Missense Mutation | CGG,TGG | R,W 465 | NP_000772.2 | |
NM_001099773.1 | 1622 | Missense Mutation | CGG,TGG | R,W 307 | NP_001093243.1 |