Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAACATTATCAGTTTGAACATGGA[C/T]CTTGAAAGGGATACGCAGAGAATAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMCO5A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMCO5A - transmembrane and coiled-coil domains 5A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152453.3 | 228 | Silent Mutation | GAC,GAT | D,D 19 | NP_689666.2 | |
XM_005254168.3 | 228 | Silent Mutation | GAC,GAT | D,D 19 | XP_005254225.1 | |
XM_011521262.2 | 228 | Silent Mutation | GAC,GAT | D,D 19 | XP_011519564.1 | |
XM_011521263.1 | 228 | Silent Mutation | GAC,GAT | D,D 19 | XP_011519565.1 | |
XM_011521264.1 | 228 | Silent Mutation | GAC,GAT | D,D 19 | XP_011519566.1 |