Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGTGACGACGCTTAGTCTGTCCGC[G/T]AACAAGATCACTGTGCTGCGGCGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614179 | ||||||||||||||||||||
Literature Links: |
ISLR2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ISLR2 - immunoglobulin superfamily containing leucine rich repeat 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130136.1 | 788 | Silent Mutation | GCG,GCT | A,A 60 | NP_001123608.1 | |
NM_001130137.1 | 788 | Silent Mutation | GCG,GCT | A,A 60 | NP_001123609.1 | |
NM_001130138.1 | 788 | Silent Mutation | GCG,GCT | A,A 60 | NP_001123610.1 | |
NM_020851.2 | 788 | Silent Mutation | GCG,GCT | A,A 60 | NP_065902.1 | |
XM_011521840.2 | 788 | Silent Mutation | GCG,GCT | A,A 60 | XP_011520142.1 | |
XM_011521841.1 | 788 | Silent Mutation | GCG,GCT | A,A 60 | XP_011520143.1 | |
XM_017022446.1 | 788 | Silent Mutation | GCG,GCT | A,A 60 | XP_016877935.1 |
LOC283731 - uncharacterized LOC283731 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |