Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGGCCCAGGAGGCCTCAAGGGCC[C/T]GGTGTGGCCTGCAAAGGAAGGTAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613948 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPATA8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPATA8 - spermatogenesis associated 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304804.1 | 270 | Missense Mutation | CCG,CTG | P,L 20 | NP_001291733.1 | |
NM_001304805.1 | 270 | Missense Mutation | CCG,CTG | P,L 20 | NP_001291734.1 | |
NM_001304806.1 | 270 | Missense Mutation | CCG,CTG | P,L 20 | NP_001291735.1 | |
NM_173499.4 | 270 | Missense Mutation | CCG,CTG | P,L 59 | NP_775770.1 | |
XM_011521265.2 | 270 | Missense Mutation | CCG,CTG | P,L 20 | XP_011519567.1 |
SPATA8-AS1 - SPATA8 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |