Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCTGAAGATGATTGGAAAATATA[C/T]TTGTTGGCCCAGAACTGTATTTGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608183 | ||||||||||||||||||||
Literature Links: |
CHSY1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHSY1 - chondroitin sulfate synthase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014918.4 | 1718 | Missense Mutation | ATA,GTA | I,V 654 | NP_055733.2 | |
XM_006720435.3 | 1718 | Missense Mutation | ATA,GTA | I,V 382 | XP_006720498.1 | |
XM_011521364.2 | 1718 | Missense Mutation | ATA,GTA | I,V 682 | XP_011519666.1 | |
XM_017022011.1 | 1718 | Missense Mutation | ATA,GTA | I,V 382 | XP_016877500.1 |