Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCCATGGTGACCAAAGTCTTCCC[A/G]CTTCTGCCACGTCCCTTCTGCCTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606266 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BMF PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BMF - Bcl2 modifying factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003940.1 | 3952 | UTR 3 | NP_001003940.1 | |||
NM_001003942.1 | 3952 | UTR 3 | NP_001003942.1 | |||
NM_001003943.2 | 3952 | UTR 3 | NP_001003943.1 | |||
NM_033503.3 | 3952 | UTR 3 | NP_277038.1 |