Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGGTCCACCTGGTGGCGGCATCCT[A/G]GCACTTCTAGCCTTCGCTGATGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612435 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLCO3A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLCO3A1 - solute carrier organic anion transporter family member 3A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145044.1 | Intron | NP_001138516.1 | ||||
NM_013272.3 | Intron | NP_037404.2 | ||||
XM_005254889.1 | Intron | XP_005254946.1 | ||||
XM_005254891.2 | Intron | XP_005254948.1 | ||||
XM_011521456.2 | Intron | XP_011519758.1 |