Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGGCCTTGATGTTCTTCAGCCAC[A/G]TGTGAATTTAGGGAGGGTTGATCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600178 MIM: 610979 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAP1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAP1A - microtubule associated protein 1A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPIP5K1 - diphosphoinositol pentakisphosphate kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130858.2 | 3801 | Silent Mutation | CAC,CAT | H,H 1203 | NP_001124330.1 | |
NM_001130859.2 | 3801 | Silent Mutation | CAC,CAT | H,H 1178 | NP_001124331.1 | |
NM_001190214.1 | 3801 | Silent Mutation | CAC,CAT | H,H 1176 | NP_001177143.1 | |
NM_014659.5 | 3801 | Silent Mutation | CAC,CAT | H,H 1178 | NP_055474.3 | |
XM_005254803.1 | 3801 | Silent Mutation | CAC,CAT | H,H 1199 | XP_005254860.1 | |
XM_005254804.1 | 3801 | Silent Mutation | CAC,CAT | H,H 1178 | XP_005254861.1 | |
XM_006720786.2 | 3801 | Silent Mutation | CAC,CAT | H,H 1136 | XP_006720849.1 | |
XM_011522249.1 | 3801 | Silent Mutation | CAC,CAT | H,H 1249 | XP_011520551.1 | |
XM_011522250.1 | 3801 | Silent Mutation | CAC,CAT | H,H 1249 | XP_011520552.1 | |
XM_011522252.1 | 3801 | Silent Mutation | CAC,CAT | H,H 1249 | XP_011520554.1 | |
XM_011522254.1 | 3801 | Silent Mutation | CAC,CAT | H,H 1249 | XP_011520556.1 | |
XM_011522258.1 | 3801 | Silent Mutation | CAC,CAT | H,H 994 | XP_011520560.1 | |
XM_011522259.1 | 3801 | Silent Mutation | CAC,CAT | H,H 1191 | XP_011520561.1 | |
XM_017022735.1 | 3801 | Silent Mutation | CAC,CAT | H,H 1249 | XP_016878224.1 | |
XM_017022736.1 | 3801 | Silent Mutation | CAC,CAT | H,H 1249 | XP_016878225.1 | |
XM_017022737.1 | 3801 | Silent Mutation | CAC,CAT | H,H 1249 | XP_016878226.1 | |
XM_017022738.1 | 3801 | Silent Mutation | CAC,CAT | H,H 1241 | XP_016878227.1 | |
XM_017022739.1 | 3801 | Silent Mutation | CAC,CAT | H,H 1228 | XP_016878228.1 | |
XM_017022740.1 | 3801 | Silent Mutation | CAC,CAT | H,H 1199 | XP_016878229.1 | |
XM_017022741.1 | 3801 | Silent Mutation | CAC,CAT | H,H 1199 | XP_016878230.1 | |
XM_017022742.1 | 3801 | Silent Mutation | CAC,CAT | H,H 1199 | XP_016878231.1 | |
XM_017022743.1 | 3801 | Silent Mutation | CAC,CAT | H,H 1199 | XP_016878232.1 | |
XM_017022744.1 | 3801 | Silent Mutation | CAC,CAT | H,H 1199 | XP_016878233.1 | |
XM_017022745.1 | 3801 | Silent Mutation | CAC,CAT | H,H 1191 | XP_016878234.1 | |
XM_017022746.1 | 3801 | Silent Mutation | CAC,CAT | H,H 1191 | XP_016878235.1 | |
XM_017022747.1 | 3801 | Silent Mutation | CAC,CAT | H,H 1186 | XP_016878236.1 | |
XM_017022748.1 | 3801 | Silent Mutation | CAC,CAT | H,H 1178 | XP_016878237.1 | |
XM_017022749.1 | 3801 | Silent Mutation | CAC,CAT | H,H 1178 | XP_016878238.1 | |
XM_017022750.1 | 3801 | Silent Mutation | CAC,CAT | H,H 1178 | XP_016878239.1 | |
XM_017022751.1 | 3801 | Silent Mutation | CAC,CAT | H,H 1178 | XP_016878240.1 | |
XM_017022752.1 | 3801 | Silent Mutation | CAC,CAT | H,H 1170 | XP_016878241.1 | |
XM_017022753.1 | 3801 | Silent Mutation | CAC,CAT | H,H 1170 | XP_016878242.1 | |
XM_017022754.1 | 3801 | Silent Mutation | CAC,CAT | H,H 1136 | XP_016878243.1 | |
XM_017022755.1 | 3801 | Silent Mutation | CAC,CAT | H,H 1136 | XP_016878244.1 | |
XM_017022756.1 | 3801 | Silent Mutation | CAC,CAT | H,H 1136 | XP_016878245.1 | |
XM_017022757.1 | 3801 | Silent Mutation | CAC,CAT | H,H 1136 | XP_016878246.1 | |
XM_017022758.1 | 3801 | Silent Mutation | CAC,CAT | H,H 923 | XP_016878247.1 | |
XM_017022759.1 | 3801 | Intron | XP_016878248.1 | |||
XM_017022760.1 | 3801 | Silent Mutation | CAC,CAT | H,H 535 | XP_016878249.1 |