Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCTGTTCTCTTCTCCATTCAAAG[C/T]AAGGTTGTGCAGGAAGAGGAGGATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606266 | ||||||||||||||||||||
Literature Links: |
BMF PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BMF - Bcl2 modifying factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003940.1 | 595 | Missense Mutation | ACT,GCT | T,A 171 | NP_001003940.1 | |
NM_001003942.1 | 595 | Missense Mutation | ACT,GCT | T,A 150 | NP_001003942.1 | |
NM_001003943.2 | 595 | Missense Mutation | TAC,TGC | Y,C 117 | NP_001003943.1 | |
NM_033503.3 | 595 | Missense Mutation | ACT,GCT | T,A 171 | NP_277038.1 |