Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCAACTGCGCCTTACACAGTACC[A/G]TGGAGGATCCTTACCAAATGTGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608986 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRTC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CRTC3 - CREB regulated transcription coactivator 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042574.2 | 333 | Missense Mutation | CAT,CGT | H,R 59 | NP_001036039.1 | |
NM_022769.4 | 333 | Missense Mutation | CAT,CGT | H,R 59 | NP_073606.3 | |
XM_005254968.2 | 333 | Missense Mutation | CAT,CGT | H,R 59 | XP_005255025.1 | |
XM_011521906.2 | 333 | Intron | XP_011520208.1 |