Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGACTTTCTCATCTAGCAACTTA[C/T]TGATTCCTTCACAAATCCTCTTGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616438 | ||||||||||||||||||||
Literature Links: |
PIRC66 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PIRC66 - piwi-interacting RNA cluster 66 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TNFAIP8L3 - TNF alpha induced protein 8 like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001311175.1 | 1050 | Missense Mutation | AAT,AGT | N,S 196 | NP_001298104.1 | |
NM_207381.3 | 1050 | Missense Mutation | AAT,AGT | N,S 284 | NP_997264.2 | |
XM_011521545.2 | 1050 | Missense Mutation | AAT,AGT | N,S 210 | XP_011519847.1 | |
XM_017022168.1 | 1050 | Missense Mutation | AAT,AGT | N,S 195 | XP_016877657.1 | |
XM_017022169.1 | 1050 | Missense Mutation | AAT,AGT | N,S 150 | XP_016877658.1 |