Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGCATAGATATTCTTCATCTACT[C/T]AGTGAAACAAATATTCTCTGGTGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEC11A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SEC11A - SEC11 homolog A, signal peptidase complex subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271918.1 | Intron | NP_001258847.1 | ||||
NM_001271919.1 | Intron | NP_001258848.1 | ||||
NM_001271920.1 | Intron | NP_001258849.1 | ||||
NM_001271921.1 | Intron | NP_001258850.1 | ||||
NM_001271922.1 | Intron | NP_001258851.1 | ||||
NM_014300.3 | Intron | NP_055115.1 |