Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTTACCTGGACACTGCTTAGAGA[A/G]TCCTCTCCAGGAATGCCACCCAGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
GABARAPL3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GABARAPL3 - GABA type A receptor associated protein like 3 pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF774 - zinc finger protein 774 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004309.2 | 239 | Missense Mutation | AAT,AGT | N,S 18 | NP_001004309.2 | |
XM_017022127.1 | 239 | UTR 5 | XP_016877616.1 | |||
XM_017022128.1 | 239 | Intron | XP_016877617.1 | |||
XM_017022129.1 | 239 | UTR 5 | XP_016877618.1 |