Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTATTTTATAAACGAGGAACTTG[A/G]GGCCCATGTAAATTAACAGACAGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 160777 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYO5A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MYO5A - myosin VA | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000259.3 | 7474 | UTR 3 | NP_000250.3 | |||
NM_001142495.1 | 7474 | UTR 3 | NP_001135967.1 | |||
XM_005254397.3 | 7474 | UTR 3 | XP_005254454.1 | |||
XM_011521606.2 | 7474 | UTR 3 | XP_011519908.2 | |||
XM_011521607.2 | 7474 | UTR 3 | XP_011519909.2 | |||
XM_011521608.2 | 7474 | UTR 3 | XP_011519910.2 | |||
XM_011521609.2 | 7474 | UTR 3 | XP_011519911.2 | |||
XM_011521610.2 | 7474 | UTR 3 | XP_011519912.2 | |||
XM_011521611.2 | 7474 | UTR 3 | XP_011519913.2 | |||
XM_011521612.2 | 7474 | UTR 3 | XP_011519914.2 | |||
XM_017022227.1 | 7474 | UTR 3 | XP_016877716.1 |