Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGTTGCAACTGAGGGAGCGCACC[A/G]TCTTCTTCTTAGTCTGCACAGACAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142704 MIM: 603247 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HDC PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HDC - histidine decarboxylase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001306146.1 | 1953 | Missense Mutation | NP_001293075.1 | |||
NM_002112.3 | 1953 | Missense Mutation | NP_002103.2 | |||
XM_017022094.1 | 1953 | Missense Mutation | XP_016877583.1 | |||
XM_017022095.1 | 1953 | Missense Mutation | XP_016877584.1 | |||
XM_017022096.1 | 1953 | Missense Mutation | XP_016877585.1 | |||
XM_017022097.1 | 1953 | Missense Mutation | XP_016877586.1 | |||
XM_017022098.1 | 1953 | Missense Mutation | XP_016877587.1 | |||
XM_017022099.1 | 1953 | Intron | XP_016877588.1 |
SLC27A2 - solute carrier family 27 member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |