Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTTCTCCAAAAAGCCAGTGGAAA[A/G]GGGGTCCTGAAGAAGTGACAAAAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606910 | ||||||||||||||||||||
Literature Links: |
BCL2L10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BCL2L10 - BCL2 like 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001306168.1 | 568 | Missense Mutation | NP_001293097.1 | |||
NM_020396.3 | 568 | Silent Mutation | NP_065129.1 |