Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAAGCTAACCAGGATTCAGGTCC[C/G]GGTTTGAGTCTTGAAGAATTTATTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602584 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RCN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RCN2 - reticulocalbin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271837.1 | 776 | Silent Mutation | CCC,CCG | P,P 185 | NP_001258766.1 | |
NM_002902.2 | 776 | Silent Mutation | CCC,CCG | P,P 167 | NP_002893.1 |