Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCGGCGGGTGGAGCCTCATCTTTC[A/G]CTGTGTATTCTCTTTGGAAAACCGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608409 | ||||||||||||||||||||
Literature Links: |
C15orf48 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C15orf48 - chromosome 15 open reading frame 48 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032413.3 | 245 | Missense Mutation | ACT,GCT | T,A 31 | NP_115789.1 | |
NM_197955.2 | 245 | Missense Mutation | ACT,GCT | T,A 31 | NP_922946.1 |
MIR147B - microRNA 147b | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPATA5L1 - spermatogenesis associated 5 like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |