Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTTTTTATCTTTGTGATTGTTGG[C/T]AGAGTTGGTTTTCCTTTGGAGGAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612820 MIM: 615176 | ||||||||||||||||||||
Literature Links: |
NPTN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NPTN - neuroplastin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161363.1 | 1335 | Missense Mutation | ACC,GCC | T,A 380 | NP_001154835.1 | |
NM_001161364.1 | 1335 | Missense Mutation | ACC,GCC | T,A 264 | NP_001154836.1 | |
NM_012428.3 | 1335 | Missense Mutation | ACC,GCC | T,A 384 | NP_036560.1 | |
NM_017455.3 | 1335 | Missense Mutation | ACC,GCC | T,A 268 | NP_059429.1 | |
XM_011521452.2 | 1335 | Intron | XP_011519754.1 |
NPTN-IT1 - NPTN intronic transcript 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
REC114 - REC114 meiotic recombination protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |