Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGTCACCCTGCTCCTCTGCCACC[A/G]ATGCCCTGATCAGTCCAGAGGCCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 123841 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPIB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPIB - peptidylprolyl isomerase B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNX22 - sorting nexin 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024798.2 | 632 | Silent Mutation | CCA,CCG | P,P 191 | NP_079074.2 | |
XM_005254677.3 | 632 | Intron | XP_005254734.1 | |||
XM_017022581.1 | 632 | Silent Mutation | CCA,CCG | P,P 163 | XP_016878070.1 |