Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCATTATTGGATTCAGTCACAAC[A/G]GATGAAGACAGCACCAGGTTCCAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 104180 | ||||||||||||||||||||
Literature Links: |
GANC PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GANC - glucosidase alpha, neutral C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301409.1 | 470 | Silent Mutation | ACA,ACG | T,T 52 | NP_001288338.1 | |
NM_001301410.1 | 470 | Silent Mutation | ACA,ACG | T,T 52 | NP_001288339.1 | |
NM_198141.2 | 470 | Silent Mutation | ACA,ACG | T,T 52 | NP_937784.2 |
TMEM87A - transmembrane protein 87A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |