Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTATTCTCCCCACACCGGGTGCC[A/G]TCCACAGCTGCATCCAGCTTGGAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605009 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAMTS7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADAMTS7 - ADAM metallopeptidase with thrombospondin type 1 motif 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014272.4 | 1779 | Silent Mutation | GAC,GAT | D,D 512 | NP_055087.2 | |
XM_005254137.3 | 1779 | Silent Mutation | GAC,GAT | D,D 512 | XP_005254194.1 | |
XM_011521166.2 | 1779 | Intron | XP_011519468.1 |