Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGTTGAGGAGACAGCAGGGGCTG[C/G]TGTGAGGTGCAGCACCCTCCCTGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611254 MIM: 170290 | ||||||||||||||||||||
Literature Links: |
KIF7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KIF7 - kinesin family member 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLIN1 - perilipin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145311.1 | 1286 | Missense Mutation | CCA,GCA | P,A 369 | NP_001138783.1 | |
NM_002666.4 | 1286 | Missense Mutation | CCA,GCA | P,A 369 | NP_002657.3 | |
XM_005254934.4 | 1286 | Missense Mutation | CCA,GCA | P,A 369 | XP_005254991.1 |