Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACCCCTGCTCCTCACCACACAGG[A/C]TCGAGCGTTTCCAGGTCAAGGGTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611254 MIM: 613298 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIF7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIF7 - kinesin family member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198525.2 | 3919 | Missense Mutation | AGC,ATC | S,I 1281 | NP_940927.2 | |
XM_011521531.2 | 3919 | Missense Mutation | AGC,ATC | S,I 1322 | XP_011519833.1 |
TICRR - TOPBP1 interacting checkpoint and replication regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308025.1 | 3919 | Intron | NP_001294954.1 | |||
NM_152259.3 | 3919 | Intron | NP_689472.3 |