Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGCAGGGGGCGAGCATGGCCGCT[C/T]CGCGAGGTGAGCCATTGGCTGGGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 154550 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM219B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM219B - family with sequence similarity 219 member B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MPI - mannose phosphate isomerase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289155.1 | 73 | Missense Mutation | CCG,TCG | P,S 4 | NP_001276084.1 | |
NM_001289156.1 | 73 | UTR 5 | NP_001276085.1 | |||
NM_001289157.1 | 73 | Missense Mutation | CCG,TCG | P,S 4 | NP_001276086.1 | |
NM_002435.2 | 73 | Missense Mutation | CCG,TCG | P,S 4 | NP_002426.1 | |
XM_011521592.1 | 73 | UTR 5 | XP_011519894.1 | |||
XM_011521593.2 | 73 | UTR 5 | XP_011519895.1 | |||
XM_017022207.1 | 73 | UTR 5 | XP_016877696.1 | |||
XM_017022208.1 | 73 | UTR 5 | XP_016877697.1 | |||
XM_017022209.1 | 73 | UTR 5 | XP_016877698.1 |