Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCAACACAAATGCATTCTGTGAC[A/G]TGTTTGAGGAGTAAATGAGGTAATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612196 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABHD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABHD2 - abhydrolase domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007011.7 | Intron | NP_008942.3 | ||||
NM_152924.4 | Intron | NP_690888.1 | ||||
XM_005254832.1 | Intron | XP_005254889.1 | ||||
XM_005254833.1 | Intron | XP_005254890.1 | ||||
XM_011521165.1 | Intron | XP_011519467.1 |