Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGCTTGAGATTCACGAGCAACTC[A/G]CAAAGGTAAGCTTGAAGGACACAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600839 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTXN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CTXN2 - cortexin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC12A1 - solute carrier family 12 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000338.2 | 631 | Missense Mutation | ACA,GCA | T,A 139 | NP_000329.2 | |
NM_001184832.1 | 631 | Missense Mutation | ACA,GCA | T,A 139 | NP_001171761.1 | |
XM_005254606.1 | 631 | Missense Mutation | ACA,GCA | T,A 139 | XP_005254663.1 |