Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTTGTGGGCAATAAGAGTGATCT[A/G]CGTCATCTCAGGGCAGTTCCTACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605570 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAB11A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RAB11A - RAB11A, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206836.1 | 511 | Silent Mutation | CTA,CTG | L,L 128 | NP_001193765.1 | |
NM_004663.4 | 511 | Silent Mutation | CTA,CTG | L,L 128 | NP_004654.1 |