Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCCACAGCTGCCCCCTGGAGGAC[C/T]CTTCCAGCTCTTCAGGACCCCCACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607503 | ||||||||||||||||||||
Literature Links: |
DISP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DISP2 - dispatched RND transporter family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033510.2 | 2454 | Missense Mutation | CCT,TCT | P,S 66 | NP_277045.1 | |
XM_011522125.2 | 2454 | Missense Mutation | CCT,TCT | P,S 66 | XP_011520427.1 | |
XM_017022687.1 | 2454 | Missense Mutation | CCT,TCT | P,S 66 | XP_016878176.1 |
PHGR1 - proline, histidine and glycine rich 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |