Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTTCGCTGGCCCGACCTGCAGCA[C/T]GCCGTGGAGCTGAAGCCCCTGTGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602931 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SMAD6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SMAD6 - SMAD family member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005585.4 | 1634 | Silent Mutation | CAC,CAT | H,H 237 | NP_005576.3 | |
XM_011521561.2 | 1634 | Intron | XP_011519863.1 |