Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCAGCCACTGTGCCCGGCCTATT[A/G]CTGCAGTTTTAAAATAATACATTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605840 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF111 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RNF111 - ring finger protein 111 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270528.1 | Intron | NP_001257457.1 | ||||
NM_001270529.1 | Intron | NP_001257458.1 | ||||
NM_001270530.1 | Intron | NP_001257459.1 | ||||
NM_017610.7 | Intron | NP_060080.6 | ||||
XM_005254479.3 | Intron | XP_005254536.1 | ||||
XM_006720575.3 | Intron | XP_006720638.1 | ||||
XM_006720579.3 | Intron | XP_006720642.1 | ||||
XM_011521694.2 | Intron | XP_011519996.2 | ||||
XM_011521696.2 | Intron | XP_011519998.2 | ||||
XM_011521697.2 | Intron | XP_011519999.2 | ||||
XM_011521699.2 | Intron | XP_011520001.2 | ||||
XM_017022336.1 | Intron | XP_016877825.1 | ||||
XM_017022337.1 | Intron | XP_016877826.1 | ||||
XM_017022338.1 | Intron | XP_016877827.1 | ||||
XM_017022339.1 | Intron | XP_016877828.1 | ||||
XM_017022340.1 | Intron | XP_016877829.1 | ||||
XM_017022341.1 | Intron | XP_016877830.1 | ||||
XM_017022342.1 | Intron | XP_016877831.1 | ||||
XM_017022343.1 | Intron | XP_016877832.1 |