Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGATTCAGAGTGTGTTGCCATGGC[A/G]GTGAGTCTAAGGAAGGTGTTCTCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607089 MIM: 177070 | ||||||||||||||||||||
Literature Links: |
CCNDBP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCNDBP1 - cyclin D1 binding protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EPB42 - erythrocyte membrane protein band 4.2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000119.2 | 1727 | Silent Mutation | ACC,ACT | T,T 595 | NP_000110.2 | |
NM_001114134.1 | 1727 | Silent Mutation | ACC,ACT | T,T 565 | NP_001107606.1 | |
XM_005254225.1 | 1727 | Silent Mutation | ACC,ACT | T,T 530 | XP_005254282.1 | |
XM_011521349.2 | 1727 | Silent Mutation | ACC,ACT | T,T 595 | XP_011519651.1 | |
XM_011521350.2 | 1727 | Silent Mutation | ACC,ACT | T,T 595 | XP_011519652.1 | |
XM_011521351.2 | 1727 | Silent Mutation | ACC,ACT | T,T 595 | XP_011519653.1 | |
XM_011521352.2 | 1727 | Silent Mutation | ACC,ACT | T,T 583 | XP_011519654.1 | |
XM_011521353.2 | 1727 | Silent Mutation | ACC,ACT | T,T 560 | XP_011519655.1 | |
XM_011521354.2 | 1727 | Silent Mutation | ACC,ACT | T,T 410 | XP_011519656.1 |