Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTCCTCTTCAGGTCATCCCCACA[C/T]AGCCTGGTCTGAAAGCCCATTTCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606758 MIM: 612771 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DUOX1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DUOX1 - dual oxidase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017434.4 | Intron | NP_059130.2 | ||||
NM_175940.2 | Intron | NP_787954.1 | ||||
XM_011521681.2 | Intron | XP_011519983.1 | ||||
XM_011521682.1 | Intron | XP_011519984.1 |
DUOXA1 - dual oxidase maturation factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276264.1 | Intron | NP_001263193.1 | ||||
NM_001276265.1 | Intron | NP_001263194.1 | ||||
NM_001276266.1 | Intron | NP_001263195.1 | ||||
NM_001276267.1 | Intron | NP_001263196.1 | ||||
NM_001276268.1 | Intron | NP_001263197.1 | ||||
NM_144565.3 | Intron | NP_653166.2 | ||||
XM_006720743.3 | Intron | XP_006720806.1 | ||||
XM_006720745.1 | Intron | XP_006720808.1 | ||||
XM_006720746.1 | Intron | XP_006720809.1 | ||||
XM_006720747.3 | Intron | XP_006720810.1 | ||||
XM_006720749.1 | Intron | XP_006720812.1 | ||||
XM_006720751.3 | Intron | XP_006720814.1 | ||||
XM_006720752.3 | Intron | XP_006720815.2 | ||||
XM_006720754.1 | Intron | XP_006720817.1 | ||||
XM_006720755.1 | Intron | XP_006720818.1 | ||||
XM_011522181.2 | Intron | XP_011520483.1 | ||||
XM_011522182.1 | Intron | XP_011520484.1 | ||||
XM_011522183.2 | Intron | XP_011520485.1 | ||||
XM_011522184.1 | Intron | XP_011520486.1 | ||||
XM_011522185.1 | Intron | XP_011520487.1 | ||||
XM_011522186.2 | Intron | XP_011520488.1 |