Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATACTGTGGAGGCTGTTGCCTATG[C/T]GCCAAAGAAGGAGCTAATAAATATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 179617 | ||||||||||||||||||||
Literature Links: |
RAD51 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RAD51 - RAD51 recombinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164269.1 | 213 | Missense Mutation | GCG,GTG | A,V 55 | NP_001157741.1 | |
NM_001164270.1 | 213 | Missense Mutation | GCG,GTG | A,V 55 | NP_001157742.1 | |
NM_002875.4 | 213 | Missense Mutation | GCG,GTG | A,V 55 | NP_002866.2 | |
NM_133487.3 | 213 | Missense Mutation | GCG,GTG | A,V 55 | NP_597994.3 | |
XM_006720626.3 | 213 | Missense Mutation | GCG,GTG | A,V 55 | XP_006720689.1 | |
XM_011521857.2 | 213 | Missense Mutation | GCG,GTG | A,V 55 | XP_011520159.1 | |
XM_011521858.2 | 213 | Missense Mutation | GCG,GTG | A,V 55 | XP_011520160.1 | |
XM_011521859.2 | 213 | Missense Mutation | GCG,GTG | A,V 55 | XP_011520161.1 | |
XM_011521860.2 | 213 | Missense Mutation | GCG,GTG | A,V 55 | XP_011520162.1 | |
XM_011521861.2 | 213 | Missense Mutation | GCG,GTG | A,V 55 | XP_011520163.1 | |
XM_011521862.2 | 213 | UTR 5 | XP_011520164.1 |
RAD51-AS1 - RAD51 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |