Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAACACTGGCCGGGAAAAACATCA[A/G]GCTTTCACCTCACTCATGGCCTCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603484 MIM: 608552 | ||||||||||||||||||||
Literature Links: |
PRC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PRC1 - protein regulator of cytokinesis 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VPS33B - VPS33B, late endosome and lysosome associated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289148.1 | 2142 | Silent Mutation | GCC,GCT | A,A 590 | NP_001276077.1 | |
NM_001289149.1 | 2142 | Silent Mutation | GCC,GCT | A,A 526 | NP_001276078.1 | |
NM_018668.4 | 2142 | Silent Mutation | GCC,GCT | A,A 617 | NP_061138.3 | |
XM_005254887.1 | 2142 | Silent Mutation | GCC,GCT | A,A 526 | XP_005254944.1 | |
XM_011521448.1 | 2142 | Silent Mutation | GCC,GCT | A,A 526 | XP_011519750.1 | |
XM_011521449.2 | 2142 | Silent Mutation | GCC,GCT | A,A 509 | XP_011519751.1 | |
XM_017022075.1 | 2142 | Silent Mutation | GCC,GCT | A,A 502 | XP_016877564.1 | |
XM_017022076.1 | 2142 | Silent Mutation | GCC,GCT | A,A 502 | XP_016877565.1 |