Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTCCCCACACCGGGTGCCGTCCA[C/T]AGCTGCATCCAGCTTGGAGTGACAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605009 | ||||||||||||||||||||
Literature Links: |
ADAMTS7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADAMTS7 - ADAM metallopeptidase with thrombospondin type 1 motif 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014272.4 | 1774 | Missense Mutation | ATG,GTG | M,V 511 | NP_055087.2 | |
XM_005254137.3 | 1774 | Missense Mutation | ATG,GTG | M,V 511 | XP_005254194.1 | |
XM_011521166.2 | 1774 | Intron | XP_011519468.1 |